To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD274 (PD-L1, B7-H1) (10F.9G2)
- Clonality:Monoclonal
- Clone:10F.9G2
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CD274
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:PDL1|B7H1
- Cat. no.:75842-364
- Supplier No.:26812-25-500
Specifications
About this item
The 10F.9G2 monoclonal antibody specifically reacts with mouse CD274, also known as B7-H1 or PD-L1, a 43 kDa glycoprotein of the B7 family of the immunoglobulin superfamily. CD274 is expressed on the B and T lymphocytes, natural killer cells, and dendritic cells. The receptor for the B7-H1 molecule is PD-1, which contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM), and is expressed on activated B and T cells. The interaction between CD274 and PD-1 seems to downregulate the T and B immune responses.
Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 10F.9G2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse