To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD27 molecule
- Clonality:Monoclonal
- Clone:LG.3A10
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CD27
- Flow cytometry:Yes
- Size:50 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Tp55|S152|T14|TNFRSF7|S152. LPFS2
- Cat. no.:75842-014
- Supplier No.:09321-25-50
Specifications
About this item
The LG.3A10 monoclonal antibody binds to CD27, 45kDA molecule in the Tumor Necrosis Factor Receptor (TNFR) superfamily. It is expressed on memory B cells, NK cells, mature T cells, and a subset of thymocytes. Cd27 is involved in the interactions between B and T cells through its ligand CD70. The LG.3A10 antibody is cross-reactive with human, mouse and rat CD27.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LG.3A10
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human