To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD27 molecule
- Clonality:Monoclonal
- Clone:O323
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD27
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:Tp55|S152|T14|TNFRSF7|S152. LPFS2
- Cat. no.:75842-010
- Supplier No.:09311-87-100
Specifications
About this item
The O323 monoclonal antibody specifically binds to the human CD27, which is a lymphocyte-specific member of the TNF/NGF-R family, expressed on a subset of human thymocytes and the majority of the mature T cells. CD27 is upregulated by T lymphocytes stimulation, and binds to CD70, enhancing the interaction between T and B lymphocytes. CD27 is a marker of memory B cells.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human