To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:RM134L
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:OX40L
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:OX-40 Ligand|gp34|CD134L|OX-40L|TXGP1|CD252|Tumor necrosis factor ligand superfamily member 4
- Cat. no.:75843-218
- Supplier No.:24611-80-25
Specifications
About this item
The RM134L monoclonal antibody specifically reacts with mouse CD252, a TNF/NGF superfamily member present on activated B lymphocytes and antigen-presenting cells. On activated B cells it enhances immunoglobulin secretion and cell proliferation. CD252 is also known as the OX-40 ligand and interacts with the OX-40 antigen found on the surface of activated T cells. The RML134L is reported to block the costimulatory activity of OX-40L.
Type: Primary
Antigen: OX40L
Clonality: Monoclonal
Clone: RM134L
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse