To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin-2 receptor alpha chain
- Clonality:Monoclonal
- Clone:BC96
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:IL2RA
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:IMD41|TCGFR|p55|interleukin 2 receptor subunit alpha|IL2R|IDDM10|CD25|IL2RA
- Cat. no.:75841-916
- Supplier No.:07311-60-100
Specifications
About this item
The BC96 monoclonal antibody specifically reacts with the 55 kDa type I transmembrane glycoprotein known as the interleukin-2 receptor α (IL-2R α, also known as CD25). CD25 is expressed by the early progenitors of T and B lymphocytes lineage, and by activated mature T and B lymphocytes. CD25 is a low affinity interleukin-2 receptor, but its association with the IL-2 receptor β chain (CD122) and the common γ chain (CD 132) results in a high affinity IL-2R complex. CD25 plays an important role in B and T cell proliferation, differentiation, and activation.
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: BC96
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human