To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin-2 receptor alpha chain
- Clonality:Monoclonal
- Clone:PC61.5
- Host:Rat
- Isotype:IgG lambda
- Reactivity:Mouse
- Antigen symbol:IL2RA
- Conjugation:BG Violet 450
- Flow cytometry:Yes
- Size:25 ug
- Form:liquid
- Antigen synonyms:IMD41|TCGFR|p55|interleukin 2 receptor subunit alpha|IL2R|IDDM10|CD25|IL2RA
- Cat. no.:75841-940
- Supplier No.:07312-40-25
Specifications
About this item
The PC61.5 antibody specifically reacts with mouse CD25, the 55 kDa low-affinity Interleukin-2 Receptor α chain (IL-2R α), expressed on early progenitors of T and B lineage, and on B and T cells. Together with CD122 (IL-2 Receptor β) and CD 132 (IL-2 Receptor γc, the common gamma chain), CD25 forms high-affinity receptor complexes for IL-2. Resting B and T cells and natural killer cells do not express IL-2Rα. Cd25 is also expressed on the dendritic cells, and it enhances lymphocyte differentiation and activation.The PC61.5 antibody blocks the binding of IL-2 to both high-affinity and low-affinity receptors.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: PC61.5
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG1, lambda
Reactivity: Mouse