To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:M1/69
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CD24
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Heat Stable Antigen|HSA
- Cat. no.:75841-776
- Supplier No.:06312-25-500
Specifications
About this item
The M1/69 monoclonal antibody specifically reacts with mouse CD24, variably glycosylated GPI-anchored membrane protein. CD24 is known as the Heat Stable Antigen (HAS or HsAG) and is found on epithelial cells, thymocytes, monocytes, erythrocytes, granulocytes, dendritic cells and lymphocytes to the exclusion of NK cells. It is useful in delineating the stages of lymphocyte development and is involved in the costimulation of CD4+ T cells by B cells. CD62P (P-Selectin) is the reported ligand of CD24.
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: M1/69
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse