To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:SN3
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD24
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Cat. no.:75841-768
- Supplier No.:06311-50-100
Specifications
About this item
The SN3 monoclonal antibody specifically reacts with human CD24, a 35-45 kDA molecule also known as the Heat Stable Antigen (HAS), Ly-52, Nectadrin. It can be used as a marker for distinguishing between lymphocyte developmental stages as its expression varies on T and B cells during differentiation. CD24 is also expressed on monocytes, dendritic cells, hematopoietic stem cells, epidermal Lagershans cells, and neurons.
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: SN3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human