To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HB5
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:CD21
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:Epstein-Barr virus receptor|Complement receptor type 2|CR2|EBV-R
- Cat. no.:75842-838
- Supplier No.:03311-50-100
Specifications
About this item
The HB5 monoclonal antibody specifically reacts with human CD21, a 145 kDA glycosylated type I integral membrane protein. CD21 is reported to be involved in B cell activation and proliferation. It is expressed on follicular dendritic cells, mature B cells, a subset of T cells, and a subset of epithelial cells. It is the receptor utilized by the Epstein-Barr virus to infect B cells.
Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: HB5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: mouse
Isotype: IgG2a, kappa
Reactivity: Human