To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B-lymphocyte antigen CD20
- Clonality:Monoclonal
- Clone:2H7
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:CD20
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:CVID5|MS4A1|Bp35|membrane spanning 4-domains A1|S7|LEU-16|MS4A2|CD20|B1
- Cat. no.:75842-738
- Supplier No.:02311-60-100
Specifications
About this item
The 2H7 monoclonal antibody specifically reacts with human CD20, a 33-37kDA B-lymphocyte surface molecule. CD20 is an unglycosylated four-transmembrane phosphoprotein expressed by B cells in all stage of development, except the final plasma cells. It has been reported that the molecule is involved in B cell activation and has also been observed on a subset of circulating T lymphocytes.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human