To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of differentiation 2
- Clonality:Monoclonal
- Clone:RPA-2.10
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD2
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:Ly-37|T11|LFA-2|SRBC-R
- Cat. no.:75843-784
- Supplier No.:04111-60-100
Specifications
About this item
The RPA-2.10 monoclonal antibody specifically binds to human CD2, a 50 kDA type I transmembrane glycoprotein. CD2 is expressed on NK cells, thymocytes, T lymphocytes, and B cell subsets. CD2 is involved in cell-cell adhesion, T cell activation, and T cell signaling. Its ligand is CD58 and it is reported to bind to CD48, CD59, and CD15. The RPA-2.10 antibody is reported to be cross-reactive with pigs and non-human primates.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human