To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of differentiation 2
- Clonality:Monoclonal
- Clone:RM2-5
- Host:Rat
- Isotype:IgG2b lambda
- Reactivity:Mouse
- Antigen symbol:CD2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Ly-37|T11|LFA-2
- Cat. no.:75843-794
- Supplier No.:04112-50-100
Specifications
About this item
The RM2-5 monoclonal antibody specifically binds to mouse CD2, a member of the Ig superfamily and type I transmembrane glycoprotein. CD2 is expressed on B cells, T cells, NK cells, and thymocytes and its ligand is CD48. It may be involved in immunoregulation, thymocyte maturation, and T-cell activation. The RM2-5 antibody has been reported to block CD2 mediated cell to cell adhesion.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RM2-5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b, lambda
Reactivity: Mouse