To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:1B1
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CD1d
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Ly-38|CD1.1
- Cat. no.:75841-496
- Supplier No.:03112-25-500
Specifications
About this item
The 1B1 monoclonal antibody specifically reacts with mouse CD1d, a 48kDA type I membrane glycoprotein with a structural homology to MHC class I molecules. CD1d is expressed by mouse leukocytes, epithelial cells, thymocytes, and dendritic cells and is the only molecules in the CD1 family to have been found in the mouse. The 1B1 antibody detects CD1d in association with the Beta2m molecule.
Type: Primary
Antigen: CD1d
Clonality: Monoclonal
Clone: 1B1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse