To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HIB19
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD19
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:B4 |Leu-12
- Cat. no.:75842-114
- Supplier No.:11211-77-25
Specifications
About this item
The HIB19 monoclonal antibody reacts with a human 95 kDa transmembrane glycoprotein known as CD19, which is expressed by B lymphocytes during all the developmental stages, except for the terminally differentiated plasma cells. CD19 is also expressed on follicular dendritic cells, and seems to ensure the regulation of B lymphocytes proliferation. CD19, CD21, CD81, MHC class II, and Leu13 can bind together and form a complex which associates with the B cell receptor (BCR) on the surface of B lymphocytes and facilitates the signal transduction.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human