To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HIB19
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD19
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:B4 |Leu-12
- Cat. no.:75842-154
- Supplier No.:11221-50-100
Specifications
About this item
The HIB19 monoclonal antibody specifically reacts with human CD19, a 95kDA type 1 transmembrane glycoprotein also known as B4. It is expressed on all stages of B cell development except for final plasma cells and on follicular dendritic cells. CD19 plays a role in B cell proliferation, activation, and differentiation. It functions as a BCR co-receptor with CD21 and CD81.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human