To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:C-X-C chemokine receptor type 4
- Clonality:Monoclonal
- Clone:2B11
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CXCR4
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:SDF-1 receptor|C-X-C chemokine receptor type 4|CXCR4|Fusin|CD184
- Cat. no.:75842-318
- Supplier No.:16912-60-100
Specifications
About this item
The 2B11 monoclonal antibody specifically reacts with mouse CD184 (CXCR4or Fusin), a seven-transmembrane G-protein-coupled receptor. It is expressed by astrocytes, B cells, T cells, monocytes, neutrophils, neurons, and hematopoietic cell types. CD184 is a coreceptor for X4 HIV-1.The monoclonal 2B11 antibody recognizes the amino terminus of CD184 and has functional activity.
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 2B11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse