To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD172a
- Clonality:Monoclonal
- Clone:P84
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:CD172a
- Flow cytometry:Yes
- Size:500ug
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:PTPNS1|SHPS-1|P84|BIT
- Cat. no.:75842-256
- Supplier No.:14812-25-500
Specifications
About this item
The P84 monoclonal antibody specifically reacts with mouse CD172a, a type I transmembrane protein otherwise known as the signal-regulatory protein alpha. It is an adhesion molecules expressed on dendritic cells, monocytes, macrophages, and neuronal tissue. It is reported to be involved in regulating the phagocytic activity of macrophages and mediating neural synaptic activity. The P84 antibody is reported to have neutralizing capability.
Type: Primary
Antigen: CD172a
Clonality: Monoclonal
Clone: P84
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse