To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Killer cell lectin-like receptor subfamily B, member 1
- Clonality:Monoclonal
- Clone:HP-3G10
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD161
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:NKRP1A|NKR-P1A
- Cat. no.:75844-368
- Supplier No.:13711-50-100
Specifications
About this item
The HP-3G10 monoclonal antibody specifically binds to an 80 kDa homodimer type II membrane glycoprotein from the C-type lectin superfamily, known as the human CD161 or NKR-P1A. CD161 is expressed on most natural killer cells, subsets of CD4+ and CD8+ T lymphocytes, γδ TCR T lymphocytes, a subset of CD3+ thymocytes, and especially on CD45RO+ T lymphocytes. Reports indicate that it may serve as a specific receptor for some natural killer cell targets and a possible stimulatory role.
Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human