To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:2.4G2
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Antigen symbol:CD16 /CD32
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:IGFR2|FCGR3|FCGR2|FC Receptor|IGFR3
- Cat. no.:75844-200
- Supplier No.:08212-50-25
Specifications
About this item
The 2.4G2 monoclonal antibody specifically reacts with an epitope on the extracellular domain of the mouse CD16 (Fc γ III) and CD 32 (Fc γ II). CD16 and CD32 are low affinity receptors for the IgG Fc domain and are expressed by B lymphocytes, NK cells, kupffer cells, mast cells, monocytes, macrophages, granulocytes, immature thymocytes, neutrophils, and some activated mature T cells.The 2.4G2 antibody blocks the binding of immunoglobulins to CD16 and CD32, and possibly to Fc γ I receptor.
Type: Primary
Antigen: CD16 /CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse