To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD15
- Clonality:Monoclonal
- Clone:HI98
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human
- Antigen symbol:CD15
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:Fucosyl Transferase 4 (FUT4)|Alpha-3-Fucosyltransferase (FCT3A)
- Cat. no.:75841-904
- Supplier No.:07211-50-25
Specifications
About this item
The HI98 monoclonal antibody specifically reacts with CD15, a 220 kDa carbohydrate moiety also known as SSEA-1, Lewis 3-fucosyl-N-acetyllactosamine (3-FAL). CD15 is expressed on most granulocytes and monocytes to the exclusion of lymphocytes and basophils. It is involved in cell adhesion, chemotaxis, and phagocytosis.
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: HI98
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human