To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD144 (VE-Cadherin)
- Clonality:Monoclonal
- Clone:BV13
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:CD144 (VE-Cadherin)
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Cdh5|Vascular endothelial cadherin|Cadherin 5
- Cat. no.:75842-286
- Supplier No.:16512-60-100
Specifications
About this item
The BV13 monoclonal antibody specifically reacts with mouse CD144 (VE-Cadherin), a 120 kDA type II Cadherin. CD144 is a hemophilic adhesion molecule of the endothelial cell line essential to maintaining cell layer integrity. It is involved in cell growth inhibition, migration, and survival.
Type: Primary
Antigen: CD144 (VE-Cadherin)
Clonality: Monoclonal
Clone: BV13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse