To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD144 (VE-Cadherin)
- Clonality:Monoclonal
- Clone:16B1
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD144 (VE-Cadherin)
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:Cadherin-5|VE-Cadherin
- Cat. no.:75842-278
- Supplier No.:16511-60-25
Specifications
About this item
The 16B1 monoclonal antibody specifically reacts with human CD144, the 140 kDA molecule called vascular endothelial (VE-cadherin) or cadherin 5. CD144 is an endothelial specific calcium-dependent adhesion molecule involved in cell contact-dependent growth inhibition, migration, survival, adhesion. It is concentrated at the intercellular boundaries of endothelial cells and essential in maintaining cell layer integrity.
Type: Primary
Antigen: CD144 (VE-Cadherin)
Clonality: Monoclonal
Clone: 16B1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human