To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD140b
- Clonality:Monoclonal
- Clone:APB5
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:CD140b
- Flow cytometry:Yes
- Size:100ug
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:PDGF receptor-b|Platelet Derived Growth Factor Receptor
- Cat. no.:75842-216
- Supplier No.:12522-25-100
Specifications
About this item
The APB5 monoclonal antibody specifically binds to the mouse CD140b molecule, otherwise known as the Platelet-Derived Growth Factor (PDGF) Receptor beta chain. CD140b is a receptor tyrosine kinase expressed on embryonic tissue and adult mesenchymal-derived cells. The APB5 antibody is reported to block the binding of PDGF-BB to the PDGF receptor.
Type: Primary
Antigen: CD140b
Clonality: Monoclonal
Clone: APB5
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse