To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:61D3
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD14
- Flow cytometry:Yes
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:LFA-1 alpha|ITGAL|LFA1A|Lymphocyte function-associated antigen 1 alpha chain
- Cat. no.:75841-738
- Supplier No.:06211-25-100
Specifications
About this item
The 61D3 monclonal antibody specifically binds to human CD14, a co-receptor for bacterial lipopolysaccharides (LPS). It is expressed on some dendritic cells, monocytes, and neutrophils. CD14 is involved in the body’s innate immune response and cytokine expression in monocytes and neutrophils.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human