To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:Sa2-8
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:CD14
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:50 μg
- Form:liquid
- Cat. no.:75841-764
- Supplier No.:06212-50-50
Specifications
About this item
The Sa2-8 monoclonal antibody specifically reacts with mouse CD14, a cell surface anchored glycoprotein that is primarily expressed on macrophages and is secreted in certain conditions. CD14 associates with LPS and LBP to form a receptor complex, which signals specifically in response to bacterial lipopolysaccharide (LPS) binding. The Sa2-8 antibody exhibits a weak antagonistic activate in relation to TNF alpha production and NF-kappa B activation.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: Sa2-8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse