To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD138 (DL-101)
- Clonality:Monoclonal
- Clone:DL-101
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD138 (DL-101)
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:B-B4|Syndecan1
- Cat. no.:75843-180
- Supplier No.:20411-60-25
Specifications
About this item
The DL-101 monoclonal antibody specifically reacts with human CD138, a 85-92 kDA type I heparan sulfate proteoglycan. CD138 is known as Syndecan-1 and is expressed on pre-B cells, immature B cells, plasma cells, and malignant plasma cells, to the exclusion of mature circulating B cells. Syndecan-1 is also expressed on vascular smooth muscle, endothelial, neural, and embryonic mesenchymal cells. It is involved in cellular proliferation, cellular migration, and acts as an extracellular matrix receptor. Among the hematopoietic elements, CD138 is a useful marker for plasma cells.
Type: Primary
Antigen: CD138 (DL-101)
Clonality: Monoclonal
Clone: DL-101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human