To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alanyl Aminopeptidase
- Clonality:Monoclonal
- Clone:WM15
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD13
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:gp150|APN|Alanyl Aminopeptidase (ANPEP)|Aminopeptidase N (PEPN)
- Cat. no.:75843-908
- Supplier No.:05211-80-100
Specifications
About this item
The WM-15 monoclonal antibody specifically reacts with human CD13, 150-170 type II transmembrane ectoenzyme also named aminopeptidase N and gp150. It is expressed by granulocytes and monocytes, epithelial cells, and endothelial cells. CD13 plays a role in the terminal degradation of peptides, antigen processing, and chemokine cleavage, and cellular adhesion.
Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: WM15
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human