To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin-3 Receptor alpha
- Clonality:Monoclonal
- Clone:6H6
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD123
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:IL-3Ra|Interleukin-3 Receptor alpha
- Cat. no.:75842-270
- Supplier No.:15311-60-25
Specifications
About this item
The 6H6 monoclonal antibody specifically reacts with human CD123, the alpha chain subunit of the IL-3 receptor. CD123’s affinity to IL-3 is low, but increases when associated with CD131. It can be found on basophils, dendritic cells, macrophages, and mast cells.
Type: Primary
Antigen: CD123
Clonality: Monoclonal
Clone: 6H6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human