To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:IL-2 Receptor beta
- Clonality:Monoclonal
- Clone:TM-beta1
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Antigen symbol:CD122
- Flow cytometry:Yes
- Size:100ug
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:IL-2Rb|IL-2 Receptor beta chain
- Cat. no.:75844-384
- Supplier No.:14312-25-100
Specifications
About this item
The TM-beta1 monoclonal antibody specifically reacts with mouse CD122, a beta chain shared by the IL-2 and IL-15 receptors. It is expressed by B cells, T cells, and NK cells and can be used as a marker for NK-cell progenitors. CD122 forms complexes involved in immunoregulation and signal transduction. The TM-beta1 antibody is reported to block the binding of IL-2 and IL-15 to the IL-2 beta receptor.
Type: Primary
Antigen: CD122
Clonality: Monoclonal
Clone: TM-beta1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse