To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin, alpha X
- Clonality:Monoclonal
- Clone:3.9
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD11c
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:ITGAX|CR4|p150
- Cat. no.:75842-820
- Supplier No.:03231-50-25
Specifications
About this item
The 3.9 monoclonal antibody specifically binds to the human adhesion glycoprotein CD11c, a 150 kDA integrin α chain also known as integrin alpha X. It is expressed on macrophages, granulocytes, monocytes, dendritic cells, natural killer cells, and subsets of B and T lymphocytes. The CD11c/CD18 complex associates with the iC3b, fibrinogen and ICAM-1 and has an important function in leukocyte adhesion.
Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: 3.9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human