To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:ICRF44
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD11b
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:C3biR|Mo1|CR3|Mac-1|ITGAM
- Cat. no.:75841-502
- Supplier No.:03211-50-25
Specifications
About this item
The ICRF44 monoclonal antibody specifically reacts with the 165 kDa human adhesion glycoprotein CD11b, which forms, together with the 95 kDa CD18 (integrin β2) a complex known as Mac-1. CD11b is expressed on the surface of activated lymphocytes, a subset of natural killer cells, granulocytes, and monocytes. It functions as a receptor in cell-cell and cell-matrix interactions and as a receptor for iC3b, ICAM-1, ICAM-2, and ICAM-3 intercellular adhesion molecules.
Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: ICRF44
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human