To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:M1/70
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:CD11b
- Conjugation:Biotin
- Flow cytometry:Yes
- Size:500 μg
- Form:liquid
- Antigen synonyms:C3biR|Mo1|CR3|Mac-1|Ly-40|ITGAM
- Cat. no.:75841-564
- Supplier No.:03221-30-500
Specifications
About this item
The M1/70 monoclonal antibody specifically reacts with the 170 kDa αM integrin chain of mouse CD11b from the Mac-1 integrin (CD11b/CD18). Mac-1 binds to C3bi, CD54 (ICAM-1), and fibrinogen, and it is expressed by granulocytes, macrophages, NK cells, myeloid-derived dendritic cells, microglia, activated lymphocytes, and mouse B-1 cells. The expression is up-regulated on activated neutrophils at the same time that L-selectin is shed from the cell surface. The M1/70 antibody is used for the detection of monocytes, granulocytes, and a subset of natural killer cells in human peripheral blood.M1/70 blocks C3bi binding and cell adherence, but not cell-mediated lysis and it cross-reacts with human CD11b.
Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Human