To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Colony stimulating factor 2 receptor alpha
- Clonality:Monoclonal
- Clone:4H1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CSF2RA
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:CSF2RA|GM-CSFR alpha chain
- Cat. no.:75844-408
- Supplier No.:18211-60-100
Specifications
About this item
The 4H1 monoclonal antibody specifically bind to human CD116, a 70-85 kD alpha chain of the GM-CSF receptor. It forms the high affinity GM-CSF receptor with CD131 as the beta chain. CD116 is expressed on macrophages, monocytes, eosinophils, dendritic cells, fibroblasts, neutrophils, and some tumor cells.
Type: Primary
Antigen: CSF2RA
Clonality: Monoclonal
Clone: 4H1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human