To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Colony stimulating factor 1 receptor
- Clonality:Monoclonal
- Clone:AFS98
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:CSF1R
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:CSF-1-R|Colony-Stimulating Factor 1 Receptor|c-fms|FMS|CSF1R
- Cat. no.:75842-334
- Supplier No.:17212-50-100
Specifications
About this item
The AFS98 monoclonal antibody specifically reacts with the mouse CD115 molecule, a 150 kDa receptor for the colony stimulating factor (CSF-1) or macrophage CFS (M-CFS), expressed by some epithelial cells, monocytes, osteoclasts, and macrophages. The colony stimulating factor-1 regulates the proliferation and the differentiation of the monocytic lineage cells. The AFS98 antibody can be used to identify myeloid lineage cells.
Type: Primary
Antigen: CSF1R
Clonality: Monoclonal
Clone: AFS98
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse