To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Endoglin
- Clonality:Monoclonal
- Clone:MJ7/18
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:ENG
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:200 μg
- Form:liquid
- Antigen synonyms:AI662476|AI528660|CD105|endoglin|END|Endo|ORW1|HHT1|S-endoglin
- Cat. no.:75842-220
- Supplier No.:12612-60-200
Specifications
About this item
The MJ7/18 monoclonal antibody specifically reacts with mouse CD105 (Endoglin), a 90kDA homodimeric glycoprotein expressed on vascular endothelial cells. CD105 is a marker for tumor angiogenesis research by identifying proliferating endothelium. It is also suggested to be involved in embryonic angiogenesis and cellular adhesion.
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: MJ7/18
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse