To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:2E7
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:CD103
- Flow cytometry:Yes
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:ITGAE
- Cat. no.:75842-258
- Supplier No.:15112-25-100
Specifications
About this item
The 2E7 monoclonal antibody specifically reacts with mouse CD103, a type I transmembrane glycoprotein in the integrin family that bind to E-cadherin. CD103 is expressed on intraepithelial lymphocyte T cells, peripheral regulatory T cells, lamina propria T cells, and a subset of dendritic cells. It is reported to be involved in T cell homing and immune system regulation.
Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: 2E7
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse