To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD20 Picoband™
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CD20
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:1 to 261
- Storage temperature:–20 °C
- Immunogen:E.coli-derived human CD20 recombinant protein (Position: M1-D261). Human CD20 shares 75% amino acid (aa) sequence identity with mouse CD20.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-700
- Supplier No.:PB9050
Specifications
About this item
Polyclonal antibody for CD20/MS4A1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. CD20/MS4A1 information: Molecular Weight: 33077 MW; Subcellular Localization: Cell membrane ; Multi-pass membrane protein . Cell membrane ; Lipid-anchor ; Tissue Specificity: Expressed on B-cells.
Type: Primary
Antigen: CD20
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human