To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:15 Lipoxygenase 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ALOX15
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:LOG15|Arachidonate 15-lipoxygenase|Arachidonate omega-6 lipoxygenase|15-LOX|ALOX15
- Amino acid number:2 to 337
- Storage temperature:–20 °C
- Immunogen:E.coli-derived human ALOX15 recombinant protein (Position: G2-P337). Human ALOX15 shares 72% and 73% amino acid (aa) sequences identity with mouse and rat ALOX15, respectively.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-512
- Supplier No.:PB9073
Specifications
About this item
Polyclonal antibody for 15 LIPOXYGENASE 1/ALOX15 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. 15 LIPOXYGENASE 1/ALOX15 information: Molecular Weight: 74804 MW; Subcellular Localization: Cytoplasm, cytosol. Cell membrane; Peripheral membrane protein. Lipid droplet. Predominantly cytosolic; becomes enriched at membranes upon calcium binding. Translocates from the cytosol to the plasma membrane when stimulated by IL13/interleukin-13 and in macrophages binding apoptotic cells; Tissue Specificity: Detected in monocytes and eosinophils (at protein level). Expressed in airway epithelial cells.
Type: Primary
Antigen: ALOX151
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human