To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Cyclin D1
- Antigen symbol:CCND1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:BCL1|PRAD1 oncogene|PRAD1|BCL-1 oncogene|G1/S-specific cyclin-D1|B-cell lymphoma 1 protein|U21B31|B-cell CLL/lymphoma 1|D11S287E
- Amino acid number:19 to 37
- Sequence:DANLLNDRVLRAMLKAEET
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Cyclin D1 (19-37aa DANLLNDRVLRAMLKAEET), different from the related mouse and rat sequences by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-310
- Supplier No.:PA1245-2
Polyclonal antibody for Cyclin D1/CCND1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Cyclin D1/CCND1 information: Molecular Weight: 33729 MW; Subcellular Localization: Nucleus . Cytoplasm . Membrane . Cyclin D-CDK4 complexes accumulate at the nuclear membrane and are then translocated to the nucleus through interaction with KIP/CIP family members.
Type: Primary
Antigen: Cyclin D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat