To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Mucin 1 cell surface associated, Picoband
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Antigen symbol:MUC1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:474 to 630
- Storage temperature:–20 °C
- Immunogen:E.coli-derived mouse MUC1 recombinant protein (Position: S474-L630). Mouse MUC1 shares 87% and amino acid (aa) sequence identity with human MUC1.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-476
- Supplier No.:PB9112
Specifications
About this item
Polyclonal antibody for MUC1, MUC1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Mouse. MUC1, MUC1 information: Molecular Weight: 64539 MW; Subcellular Localization: Apical cell membrane; Single-pass type I membrane protein. Exclusively located in the apical domain of the plasma membrane of highly polarized epithelial cells. After endocytosis, internalized and recycled to the cell membrane. Located to microvilli and to the tips of long filopodial protusions (By similarity); Tissue Specificity: Expressed in a variety of epithelial tissues.
Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat