About this item
PowerForm® gloves works with complete confidence knowing that this heavy-duty extended cuff glove has wearers covered, offering excellent grip and optimal chemical resistance.
- Fully textured surface for superior wet and oily grip
- Heavy-duty chemical resistance
- Touch-screen compatible
- Powder-free to prevent contamination
- Clean processing technology helps prevent skin irritation and workplace contamination
- Optimal thickness for durability and tactile sensitivity
MegaMan® pushes the boundaries of single-use hand protection performance, with new levels of abrasion resistance, sweat management, and comfort that deliver in exceedingly severe environments. Reach for MegaMan® on the toughest jobs, when protection matters most.
Loaded with features that add to this glove's performance capability, this glove line is designed and built to the very highest standards. Featuring 14 mil fingertips combined with a 12 mil palm thickness, this single-use glove also offers a full 12" length for unmatched splash protection and the ability to provide additional coverage to lab sleeves.
For decades SW® has been the premium manufacturer of single-use performance products. With a relentless focus on new manufacturing technologies and materials that drive innovation, SW® single-use styles continuously push the boundaries of quality and worker-enhancing performance that fuels business success. The SW® difference: 100% air inspected, 75% less residue, consistent tip to cuff.
Specifications
- Catalog No:
- 76051-160
- Model:
- MM-211-120-BL
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Disposable
- Latex/Latex-free:
- Latex-free
- Powdered/Powder-free:
- Powder-free
- EN norm:
- EN 420:2003
- Food contact approved:
- Yes