Upchurch Scientific® Barbed Reducing Union, IDEX Health & Science
Catalog #: 10497-952
Supplier: Upchurch Scientific
Specifications
- Tubing I.D.:6.4 and 9.5 mm
- Cat. no.:10497-952
- Supplier No.:P-810
Specifications
About this item
Barbed Reducing Union made of PP, and it is suitable for joining two peristaltic tubes with different inner diameters together
- It effectively joins one piece of soft-walled tubing to a second piece of differently sized soft-walled tubing
Customers Who Bought This Also Bought