Specifications
- Description:ImageXpress® Nano automated imaging system for microscopy assays
- Cat. no.:CA76177-144
Specifications
About this item
Easy, automated cellular imaging and analysis. The ImageXpress® Nano Automated Imaging System enables researchers to get better data faster and improve collaborations with peers anywhere, anytime.
Automated imaging now within reach of every lab
The right sensitivity to scale up microscopy assays
Drawing upon extensive feedback from customers coupled with over 30 years of imaging expertise, the system enables researchers to capture high quality images and garner more relevant, quantitative data. The imaging system can be configured with preferred filters and objectives. Additional options include transmitted light capability for imaging of non-labeled samples and sample incubation (temperature, humidity, CO2), provided through an environmental control feature.
Acquire, share, and present data; no matter where you are
Access the system and data online with the browser-based MetaXpress® High-Content Image Acquisition and Analysis Software. Make sharing, presenting, and collaborating with peers easier with built-in sharing and remote access capabilities. The software comes with 25 predefined analysis protocols and simple, linear workflows to get users started quickly and propel research forward.
MetaXpress® High-Content Image Acquisition and Analysis Software
Streamline your image analysis with MetaXpress® High-Content Image Acquisition and Analysis Software that includes tools for 2D and 3D imaging and time lapse analysis, as well as a range of needs from ease-of-use through to proprietary assay design. MetaXpress® Software is included with the system.