Succinic acid 99.0-100.5% (by acidimetry), crystalline, EMPROVE® ESSENTIAL NF, JPE, ACS, SAFC®
Catalog #: CA1.00681.5000
Supplier: MilliporeSigma
CAS Number: 110-15-6
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle
- Size:5 kg
- Cat. no.:CA1.00681.5000
- Supplier No.:1.00681.5000
Specifications
Specification Test Results
- Assay (acidimetric):99.0 − 100.5%
- Assay (HPLC):99.0 − 100.5%
- Identity:Conforms
- Appearance of solution (5%; water):Conforms
- In water insoluble matter:≤0.01%
- Melting range (lower value):≥185.0 °C
- Melting range (upper value):≤190.0 °C
- Melting point:185.0 − 190.0 °C
- Chloride (Cl):≤0.001%
- Phosphate (PO₄):≤0.001%
- Sulfate (SO₄):≤0.003%
- Heavy metals (as Pb):≤0.0020%
- Heavy metals (as Pb) (ACS):≤5 ppm
- Nitrogen compounds (as N):≤0.001%
- Fe (Iron):≤5 ppm
- As (Arsenic):≤0.0002%
- Substances reducing permanganate:Conforms
- Residual solvents (ICH Q3C):Excluded by production process
- Sulfated ash (600 °C):≤0.02%
- Solubility:58 g/l (20 °C)
- Bulk Density:940 kg/m³
- pH:2.7 (10 g/l, H₂O, 20 °C)
- Ignition Point:470 °C
- SAFC®, Succinic acid 99.0-100.5% (by acidimetry), crystalline, EMPROVE® ESSENTIAL NF, JPE, ACS:
Specification Test Results
About this item
Synonyms: 1,4-Butanedioic acid
:Storage Temperature: Ambient
Boiling Pt:235 °C (1013 hPa)
Melting Pt:185 …190 °C
Density:1.56 g/cm³ (20 °C)
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
Flash Pt:206 °C
:Boiling Pt:
:Boiling Pt: 235 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 185 …190 °C
:Flash Pt:
:Flash Pt: 206 °C
:M.W. 118.09 g/mol
:Succinic acid
Formula:HOOCCH₂CH₂COOH
MW:118.09 g/mol
MDL Number:MFCD00002789
CAS Number:110-15-6
EINECS:203-740-4
:235 °C (1013 hPa)
:206 °C
:GHS05
Storage Temperature:Ambient
:12,09038
Customers Who Bought This Also Bought