Human Synthetic DCD-1L Trifluoroacetate
Catalog #: H-7316.0500BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Protein/peptide type:Synthetic
- Species:Human
- CAS No.:478898-18-9
- Size:0.5 mg
- Storage conditions:–20 ± 5 °C
- Protein/peptide name:DCD-1L Trifluoroacetate
- Molecular weight:4818.5 g/mol
- Sequence:SSLLEKGLDGAKKAVGGLGKLGKDAVEDLESVGKGAVHDVKDVLDSVL
- Molecular formula:C₂₁₀H₃₅₉N₅₇O₇₁
- Cat. no.:H-7316.0500BA
- Supplier No.:H-7316.0500
Specifications
About this item
The protein dermcidin expressed in the sweat glands is secreted into the sweat and transported to the epidermal surface. The concomitant processing of the protein yields a broad-spectrum antimicrobial 47 amino acid peptide, dermcidin-1 (DCD-1).
Dermcidin and naturally occurring fragments as DCD-1L do not perturb membrane permeability and bind to distinct surface molecules. It stimulates keratinocytes to generate cytokines and chemokines such as TNF-α, IL-8, CXCL10, and CCL20.
Sequence: H-Ser-Ser-Leu-Leu-Glu-Lys-Gly-Leu-Asp-Gly-Ala-Lys-Lys-Ala-Val-Gly-Gly-Leu-Gly-Lys-Leu-Gly-Lys-Asp-Ala-Val-Glu-Asp-Leu-Glu-Ser-Val-Gly-Lys-Gly-Ala-Val-His-Asp-Val-Lys-Asp-Val-Leu-Asp-Ser-Val-Leu-OH trifluoroacetate salt
Customers Who Bought This Also Bought