Specifications
- Description:EKONO* Hybridiztion Buffer
- Size:1 L
- Cat. no.:CA82021-338
- Supplier No.:786-160
Specifications
About this item
A high performance hybridization buffer system providing researchers with unprecedented simplicity, reliability, accuracy, and cost saving for hybridization
Protocol involves incubation of blots with denatured probes in EKONO™ hybridization buffer followed by washing of the blot to remove excess unhybridized probes. Easy-to-use EKONO™ provides high resolution and sensitivity.
Suitable for Southern/Northern or Southwestern hybridization analysis, hybridization with oligonucleotides or other types of probes, and for hybridization screening of gene libraries.
EKONO™ hybridization buffer is suitable for all types of probes