About this item
Floor Mounted Batch Pressure Filters are manufactured for batch filtering of slurries under pressure, up to 75 psi. They offer a quick, convenient method of dewatering slurries, in two sizes, 11.5 gallon and 3 gallon capacity.
Units feature a screw jack lid opening and closing mechanism, swing away design for filling, inspection or cleaning
Also included with each pressure filter is a filter cloth, a package of 100 high wet strength filter papers with a 25 nominal micron pore opening, internal piping with pressure relief valve, pressure gauge, bleed off valve, and drain valve. All that is required for operation is a slurry, and a source of compressed air (at least 1 CFM @ 60 psi).
The base plate with the filter grid has the same screw jack opening and closing mechanism and swing away design for easy removal of the filtered solids.