Ethyl acetate ≥99.8%, Biotechnology Grade, Burdick & Jackson™
Supplier: Burdick & Jackson
New Product
Certificates
About this item
Synonyms: Acetic acid ethyl ester
Formula:CH₃COOC₂H₅
MW:88.11 g/mol
Boiling Pt:77 °C (1.013 hPa)
Melting Pt:-84 °C
Density:0.901 g/cm³
MDL Number:MFCD00009171
CAS Number:141-78-6
:Storage Temperature: Ambient
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
Flash Pt:11.5 %(V)
:Boiling Pt:
:Boiling Pt: 77 °C (1.013 hPa)
:Melting Pt:
:Melting Pt: -84 °C
:Flash Pt:
:Flash Pt: 11.5 %(V)
:M.W. 88.11 g/mol
:Ethyl acetate
Storage Temperature:Ambient
EINECS:205-500-4
:77 °C (1.013 hPa)
UN:1173
:3
:II
:11.5 %(V)
:GHS02
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.8 %
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.01 %
- Titrable Acid:Max. 0.0009 meq/g
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 275 nm:Max. 0.01
- Absorbance at 263 nm:Max. 0.05
- Absorbance at 254 nm:Max. 1.00
- Burdick & Jackson™, Ethyl acetate ≥99.8%, Biotechnology Grade:
Specification Test Results
Customers Who Bought This Also Bought