EDTA disodium salt concentrate for dilution to 1L, 0.1 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
Catalog #: BJ38057-1EA
Supplier: Honeywell Research Chemicals
CAS Number: 139-33-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Ampoule
- Size:1 Each
- Cat. no.:BJ38057-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- Fluka™, EDTA disodium salt concentrate for dilution to 1L, 0.1 M EDTA-Na2 volumetric solution, for complexometry:
Specification Test Results
About this item
Synonyms: Idranal III,Edetic acid disodium salt,Kestranal® 2S,Ethylenedinitrilotetra-acetic acid disodium salt,Disodium dihydrogen ethylenediaminetetraacetate
:Storage Temperature: Ambient
Melting Pt:no data available
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: no data available
:Flash Pt:
:Flash Pt:
:M.W. 336.21 g/mol
:EDTA disodium salt
MW:336.21 g/mol
Storage Temperature:Ambient
:12,03556
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
CAS Number:139-33-3
:Melting Pt: no data available
Customers Who Bought This Also Bought