Specifications
- Description:Multiple FIB Grid Holder
- Cat. no.:103305-528
- Supplier No.:75950-05
Specifications
About this item
Larger grid holder for focused ion beam.
Larger size enables this FIB grid holder to handle multiple FIB grids of the same thickness
Overall dimension: 22 5 x 29 x 13.5 mm
Material: Vacuum grade aluminum with brass screws
Can also be used to safely store FIB grids with thin section (lamellae) attached.